Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3,4,5-trimethoxybenzene, 97%

Catalog No. AC458131000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458131000 100 g
AC458130050 5 g
AC458130250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC458131000 Supplier Thermo Scientific Chemicals Supplier No. 458131000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2675-79-8 | C9H11BrO3 | 247.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2675-79-8
Molecular Formula C9H11BrO3
Molecular Weight (g/mol) 247.09
MDL Number MFCD00017169
InChI Key XAOOZMATJDXDQJ-UHFFFAOYSA-N
Synonym 1-bromo-3,4,5-trimethoxybenzene, 3,4,5-trimethoxybromobenzene, bromotrimethoxybenzene, benzene, 5-bromo-1,2,3-trimethoxy, 5-bromo-1,2,3-trimethoxy-benzene, pubchem10980, 3,4,5-trimethoxybromoben, ksc490o1d, 3,4,5-trimethoxyphenylbromide, 3,4,5-triemethoxyphenylbromide
PubChem CID 75885
IUPAC Name 5-bromo-1,2,3-trimethoxybenzene
SMILES COC1=CC(Br)=CC(OC)=C1OC

Specifications

Melting Point 78.0°C to 82.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 100 g
Formula Weight 247.09
Percent Purity 97%
Chemical Name or Material 1-Bromo-3,4,5-trimethoxybenzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
EINECSNumber 220-223-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.