accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2675-94-7

IUPAC Name: N,N-diethylprop-2-enamide
Synonyms: N,N-diethylprop-2-enamide
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: OVHHHVAVHBHXAK-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C=C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results