Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Diethylacrylamide

Catalog No. 77321225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-321-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 77-321-225G Supplier Sigma Aldrich Supplier No. 77321225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acrylamide monomer used to make polymers for drug delivery; thermal responsive polymers; and solutions with specifically tuned LCSTs.

Specifications

CAS 2675-94-7
Density 0.924 (at 25°C)
Molecular Formula C7H13NO
Refractive Index n20/D 1.468
Linear Formula C7H13NO
Quantity 25 g
MDL Number MFCD00078408
Synonym N,N-Diethyl-2-propenamide; Acrylic acid diethylamide; DEAA; DEAAm; DEAM
Molecular Weight (g/mol) 127.18
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.