accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26761-12-6

IUPAC Name: 1,2,2a,3,4,5-hexahydroacenaphthylene
Synonyms: 1,2,2a,3,4,5-hexahydroacenaphthylene
Molecular Weight (g/mol): 158.24
Molecular Formula: C12H14
InChi Key: BDAGIAXQQBRORQ-UHFFFAOYNA-N
SMILES: C1CC2=CC=CC3=C2C1CCC3

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results