Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5,11-Tetrahydroacenaphthene 96.0+%, TCI America™
SDP

Catalog No. T124910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T124910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T124910G Supplier TCI America Supplier No. T124910G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26761-12-6
Molecular Formula C12H14
Molecular Weight (g/mol) 158.24
MDL Number MFCD00143054
InChI Key BDAGIAXQQBRORQ-UHFFFAOYNA-N
Synonym Tetraphthene

Specifications

Color Yellow
Quantity 10 g
Formula Weight 158.24
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4,5,11-Tetrahydroacenaphthene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.