accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONO

CAS RN 2683-66-1

IUPAC Name: 4-(benzyloxy)pyridin-1-ium-1-olate
Synonyms: 4-(benzyloxy)pyridin-1-ium-1-olate
Molecular Weight (g/mol): 201.23
Molecular Formula: C12H11NO2
InChi Key: SUSQPKJQYWTFPU-UHFFFAOYSA-N
SMILES: [O-][N+]1=CC=C(OCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results