accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 268551-65-1

IUPAC Name: tert-butyl N-carbamothioylcarbamate
Synonyms: tert-butyl N-carbamothioylcarbamate
Molecular Weight (g/mol): 176.23
Molecular Formula: C6H12N2O2S
InChi Key: JKXQTODLIZBUDE-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC(N)=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)