accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNH2CH3H3CH2N

CAS RN 2687-27-6

IUPAC Name: 4-(2-{3-[2-(4-aminophenyl)propan-2-yl]phenyl}propan-2-yl)aniline
Synonyms: 4-(2-{3-[2-(4-aminophenyl)propan-2-yl]phenyl}propan-2-yl)aniline
Molecular Weight (g/mol): 344.5
Molecular Formula: C24H28N2
InChi Key: KWOIWTRRPFHBSI-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC=C(N)C=C1)C1=CC(=CC=C1)C(C)(C)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results