Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis[2-(4-aminophenyl)-2-propyl]benzene 98.0+%, TCI America™
SDP

Catalog No. B385325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B385325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B385325G Supplier TCI America Supplier No. B385325G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 2687-27-6
Molecular Formula C24H28N2
Molecular Weight (g/mol) 344.502
MDL Number MFCD00274255
InChI Key KWOIWTRRPFHBSI-UHFFFAOYSA-N
Synonym alpha,alpha′C-Bis(4-aminophenyl)-1,3-diisopropylbenzene, Bisaniline M, 4,4′C-(1,3-Phenylenediisopropylidene)bisaniline
PubChem CID 3292101
IUPAC Name 4-[2-[3-[2-(4-aminophenyl)propan-2-yl]phenyl]propan-2-yl]aniline
SMILES CC(C)(C1=CC=C(C=C1)N)C2=CC(=CC=C2)C(C)(C)C3=CC=C(C=C3)N

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 25 g
Formula Weight 344.50
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[2-(4-aminophenyl)-2-propyl]benzene
EINECSNumber (4)-1612
RTECSNumber CV6965000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.