accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHClClNH2NH2N

CAS RN 26878-35-3

IUPAC Name: dihydrogen pyridine-2,5-diamine dichloride
Synonyms: dihydrogen 2,5-diaminopyridine dichloride
Molecular Weight (g/mol): 182.05
Molecular Formula: C5H9Cl2N3
InChi Key: RWHNUWWUPZKDQP-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC1=CC=C(N)N=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)