Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diaminopyridine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. D33105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33105G 5 g
D331025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33105G Supplier TCI America Supplier No. D33105G

Chemical Identifiers

CAS 26878-35-3
Molecular Formula C5H9Cl2N3
Molecular Weight (g/mol) 182.048
InChI Key RWHNUWWUPZKDQP-UHFFFAOYSA-N
Synonym 2,5-diaminopyridine dihydrochloride, pyridine-2,5-diamine dihydrochloride, 2,5-diaminopyridine 2hcl, pyridin-2,5-diamindihydrochlorid, 2,5-diaminopyridinedihydrochloride, pubchem5497, acmc-209gtk, pyridine-2,5-diamine, chloride, chloride
PubChem CID 117889
IUPAC Name pyridine-2,5-diamine;dihydrochloride
SMILES C1=CC(=NC=C1N)N.Cl.Cl

Specifications

Melting Point 246°C
Color Purple
Quantity 5 g
Formula Weight 182.05
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diaminopyridine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.