accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26883-70-5

IUPAC Name: cyclohexane-1,3-diamine
Synonyms: 1,3-cyclohexanediamine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: GEQHKFFSPGPGLN-UHFFFAOYNA-N
SMILES: NC1CCCC(N)C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results