accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2689-37-4

IUPAC Name: N,N-diethyl-2-fluoro-4-nitroaniline
Synonyms: N,N-diethyl-2-fluoro-4-nitroaniline
Molecular Weight (g/mol): 212.22
Molecular Formula: C10H13FN2O2
InChi Key: FPVHBAPCTSCZKR-UHFFFAOYSA-N
SMILES: CCN(CC)C1=C(F)C=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results