accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2689-39-6

IUPAC Name: 4-(2-fluoro-4-nitrophenyl)morpholine
Synonyms: 4-(2-fluoro-4-nitrophenyl)morpholine
Molecular Weight (g/mol): 226.21
Molecular Formula: C10H11FN2O3
InChi Key: ZEQCFSSBZPZEFJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=C(C=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results