Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Fluoro-4-nitrophenyl)morpholine, 98%

Catalog No. AAH6194014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6194014 25 g
AAH6194006 5 g
AAH6194022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6194014 Supplier Thermo Scientific Chemicals Supplier No. H6194014
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2689-39-6 | C10H11FN2O3 | 226.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2689-39-6
Molecular Formula C10H11FN2O3
Molecular Weight (g/mol) 226.21
MDL Number MFCD03138381
InChI Key ZEQCFSSBZPZEFJ-UHFFFAOYSA-N
Synonym 4-2-fluoro-4-nitrophenyl morpholine, morpholine, 4-2-fluoro-4-nitrophenyl, 4-2-fluoro-4-nitro-phenyl-morpholine, 3-fluoro-4-morpholinonitrobenzene, acmc-209gtt, cambridge id 6614516, 3fluoro-4-morpholinonitrobenzene, 3-fluoro-4-morpholinyl nitrobenzene, 3-fluoro-4-morpholinyl-nitrobenzene, 1-fluoro-2-morpholino-5-nitrobenzene
PubChem CID 2905489
IUPAC Name 4-(2-fluoro-4-nitrophenyl)morpholine
SMILES [O-][N+](=O)C1=CC(F)=C(C=C1)N1CCOCC1

Specifications

Quantity 25 g
Formula Weight 226.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-(2-Fluoro-4-nitrophenyl)morpholine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.