accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 269055-15-4

IUPAC Name: 4-({6-amino-5-bromo-2-[(4-cyanophenyl)amino]pyrimidin-4-yl}oxy)-3,5-dimethylbenzonitrile
Synonyms: etravirine intelence C20H15brN6O
Molecular Weight (g/mol): 435.29
Molecular Formula: C20H15BrN6O
InChi Key: PYGWGZALEOIKDF-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1OC1=NC(NC2=CC=C(C=C2)C#N)=NC(N)=C1Br)C#N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results