Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Etravirine, Thermo Scientific™

Catalog No. AC468725000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468725000 500 mg
AC468721000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468725000 Supplier Thermo Scientific Chemicals Supplier No. 468725000
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 269055-15-4
Molecular Formula C20H15BrN6O
Molecular Weight (g/mol) 435.29
MDL Number MFCD09837879
InChI Key PYGWGZALEOIKDF-UHFFFAOYSA-N
Synonym 4-((6-Amino-5-bromo-2-((4-cyanophenyl)amino)pyrimidin-4-yl)oxy)-3,5-dimethylbenzonitrile
IUPAC Name 4-({6-amino-5-bromo-2-[(4-cyanophenyl)amino]pyrimidin-4-yl}oxy)-3,5-dimethylbenzonitrile
SMILES CC1=CC(=CC(C)=C1OC1=NC(NC2=CC=C(C=C2)C#N)=NC(N)=C1Br)C#N

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White to Yellow to Beige
Melting Point 262°C to 265°C
Solubility Insoluble in water
Quantity 500 mg
Formula Weight 435.28
Chemical Name or Material Etravirine
Hazard Category GHS07: Exclamation mark
ShelfLife 5 years
Recommended Storage Freezer -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.