accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONNNNNOONOONOO

CAS RN 2691-41-0

IUPAC Name: 1,3,5,7-tetranitro-1,3,5,7-tetrazocane
Synonyms: octogen HMX cyclotetramethylene tetranitramine tetramethylenetetranitramine
Molecular Weight (g/mol): 296.16
Molecular Formula: C4H8N8O8
InChi Key: UZGLIIJVICEWHF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)N1CN(CN(CN(C1)[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results