accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26944-31-0

CH3ONOO

CAS RN 26944-31-0

IUPAC Name: N,N-diformylacetamide
Synonyms: N,N-diformylacetamide
Molecular Weight (g/mol): 115.09
Molecular Formula: C4H5NO3
InChi Key: ZAHBLOGBMDVRDK-UHFFFAOYSA-N
SMILES: CC(=O)N(C=O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results