accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 2699-13-0

IUPAC Name: 3-methoxycyclohex-1-ene
Synonyms: 3-methoxycyclohex-1-ene
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: OKDKFTKUXADLSJ-UHFFFAOYNA-N
SMILES: COC1CCCC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results