accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26990-69-2

IUPAC Name: 1,3-dimethyl-5-(morpholin-4-yl)-1H-pyrazole-4-carbaldehyde
Synonyms: 1,3-dimethyl-5-(morpholin-4-yl)pyrazole-4-carbaldehyde
Molecular Weight (g/mol): 209.25
Molecular Formula: C10H15N3O2
InChi Key: LWYDVSXDKNQUNB-UHFFFAOYSA-N
SMILES: CN1N=C(C)C(C=O)=C1N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results