Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethyl-5-morpholino-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC19204DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC19204DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC19204DA Supplier Thermo Scientific Chemicals Supplier No. CC19204DA
Add to Cart

Chemical Identifiers

CAS 26990-69-2
Molecular Formula C10H15N3O2
Molecular Weight (g/mol) 209.249
MDL Number MFCD02681945
InChI Key LWYDVSXDKNQUNB-UHFFFAOYSA-N
Synonym 1,3-dimethyl-5-morpholino-1h-pyrazole-4-carbaldehyde, 1,3-dimethyl-5-morpholin-4-yl-1h-pyrazole-4-carbaldehyde, 1,3-dimethyl-5-morpholin-4-yl pyrazole-4-carbaldehyde, 1,3-dimethyl-5-morpholin-4-yl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde,1,3-dimethyl-5-4-morpholinyl
PubChem CID 2776494
IUPAC Name 1,3-dimethyl-5-morpholin-4-ylpyrazole-4-carbaldehyde
SMILES CC1=NN(C(=C1C=O)N2CCOCC2)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 209.25
Percent Purity 97%
Chemical Name or Material 1, 3-Dimethyl-5-morpholino-1H-pyrazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.