accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONOONOO

CAS RN 2702-58-1

IUPAC Name: methyl 3,5-dinitrobenzoate
Synonyms: methyl 3,5-dinitrobenzoate
Molecular Weight (g/mol): 226.14
Molecular Formula: C8H6N2O6
InChi Key: POGCCFLNFPIIGW-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results