accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOOOClCl

CAS RN 2702-72-9

IUPAC Name: sodium 2-(2,4-dichlorophenoxy)-2-oxoethan-1-olate
Synonyms: 2,4-D, sodium salt
Molecular Weight (g/mol): 243.01
Molecular Formula: C8H5Cl2NaO3
InChi Key: SFYPSBLCFRKKDT-UHFFFAOYSA-N
SMILES: [Na+].[O-]CC(=O)OC1=CC=C(Cl)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results