accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFFFFFFFFFFF

CAS RN 2708-07-8

IUPAC Name: 3,3,4,4,5,5,6,6,7,7,7-undecafluoroheptan-2-one
Synonyms: 3,3,4,4,5,5,6,6,7,7,7-undecafluoroheptan-2-one
Molecular Weight (g/mol): 312.08
Molecular Formula: C7H3F11O
InChi Key: HWGNOHFZECDUBX-UHFFFAOYSA-N
SMILES: CC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results