Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Undecafluoroamyl Ketone 98.0+%, TCI America™
SDP

Catalog No. U00715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00715G Supplier TCI America Supplier No. U00715G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2708-07-8
Molecular Formula C7H3F11O
Molecular Weight (g/mol) 312.08
MDL Number MFCD02093506
InChI Key HWGNOHFZECDUBX-UHFFFAOYSA-N
Synonym 1H,1H,1H-Undecafluoro-2-heptanone, Methyl Perfluoropentyl Ketone, 3,3,4,4,5,5,6,6,7,7,7-Undecafluoro-2-heptanone, Methyl Perfluoroamyl Ketone, Methyl Undecafluoropentyl Ketone
PubChem CID 15859812
IUPAC Name 3,3,4,4,5,5,6,6,7,7,7-undecafluoroheptan-2-one
SMILES CC(=O)C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Quantity 5 g
UN Number 1224
Formula Weight 312.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Undecafluoroamyl Ketone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.