accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COON

CAS RN 27104-73-0

IUPAC Name: methyl isoquinoline-3-carboxylate
Synonyms: methyl isoquinoline-3-carboxylate
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: ZBCGBIZQNMVMPC-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results