accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3OOH3C

CAS RN 27132-23-6

IUPAC Name: 1,3-dimethyl 2,2-diethylpropanedioate
Synonyms: 1,3-dimethyl 2,2-diethylpropanedioate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: AYBLPISRXMEMBV-UHFFFAOYSA-N
SMILES: CCC(CC)(C(=O)OC)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results