Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Diethylmalonate 98.0+%, TCI America™
SDP

Catalog No. D029225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D029225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D029225G Supplier TCI America Supplier No. D029225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27132-23-6
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00008444
InChI Key AYBLPISRXMEMBV-UHFFFAOYSA-N
Synonym Diethylmalonic Acid Dimethyl Ester
PubChem CID 117935
IUPAC Name dimethyl 2,2-diethylpropanedioate
SMILES CCC(CC)(C(=O)OC)C(=O)OC

Specifications

Color Colorless
Boiling Point 207°C
Quantity 25 g
Formula Weight 188.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Diethylmalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.