accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOS

CAS RN 27144-18-9

IUPAC Name: 3-[(triphenylmethyl)sulfanyl]propanoic acid
Synonyms: 3-[(triphenylmethyl)sulfanyl]propanoic acid
Molecular Weight (g/mol): 348.46
Molecular Formula: C22H20O2S
InChi Key: AECGEIVNZGQBJT-UHFFFAOYSA-N
SMILES: OC(=O)CCSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results