Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Tritylthio)propionic acid, 97%

Catalog No. AAH6009303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6009303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6009303 Supplier Thermo Scientific Chemicals Supplier No. H6009303
  • $149.20
    $126.65 / Each of 1

    Save $22.55

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 27144-18-9 | C22H20O2S | 348.46 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

Solubility
Partly miscible in water.

Notes
Stor
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27144-18-9
Molecular Formula C22H20O2S
Molecular Weight (g/mol) 348.46
MDL Number MFCD00237291
InChI Key AECGEIVNZGQBJT-UHFFFAOYSA-N
Synonym 3-tritylthio propanoic acid, s-trityl-3-mercaptopropionic acid, 3-tritylthio propionic acid, 3-tritylsulfanylpropionic acid, 3-triphenylmethyl sulfanyl propanoic acid, 3-tritylsulfanyl-propionic acid, 3-tritylmercapto propionic acid, s-trityl-beta-mercaptopropionic acid, propanoic acid, 3-triphenylmethyl thio, 3-tritylsulfanyl propanoic acid
PubChem CID 262767
IUPAC Name 3-tritylsulfanylpropanoic acid
SMILES OC(=O)CCSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Linear Formula (C6H5)3CSCH2CH2COOH
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Partly miscible in water.
Formula Weight 348.46
Percent Purity 97%
Chemical Name or Material 3-(Tritylthio)propionic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.