accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHO

CAS RN 27147-71-3

IUPAC Name: 2-methylhexadecanoic acid
Synonyms: 2-methylhexadecanoic acid
Molecular Weight (g/mol): 270.46
Molecular Formula: C17H34O2
InChi Key: AXPAUZGVNGEWJD-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCC(C)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results