Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylhexadecanoic Acid 98.0+%, TCI America™
SDP

Catalog No. M10261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10261G Supplier TCI America Supplier No. M10261G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27147-71-3
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00142945
InChI Key AXPAUZGVNGEWJD-UHFFFAOYSA-N
Synonym 2-Methylpalmitic Acid
PubChem CID 117384
ChEBI CHEBI:85057
IUPAC Name 2-methylhexadecanoic acid
SMILES CCCCCCCCCCCCCCC(C)C(=O)O

Specifications

Melting Point 48°C
Color White
Boiling Point 177°C
Quantity 1 g
Formula Weight 270.46
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylhexadecanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.