accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNH2CH3H3CNH2

CAS RN 2716-10-1

IUPAC Name: 4-(2-{4-[2-(4-aminophenyl)propan-2-yl]phenyl}propan-2-yl)aniline
Synonyms: 4-(2-{4-[2-(4-aminophenyl)propan-2-yl]phenyl}propan-2-yl)aniline
Molecular Weight (g/mol): 344.5
Molecular Formula: C24H28N2
InChi Key: HESXPOICBNWMPI-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC=C(N)C=C1)C1=CC=C(C=C1)C(C)(C)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results