Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha'-Bis(4-aminophenyl)-1,4-diisopropylbenzene 99.0+%, TCI America™
SDP

Catalog No. B1562500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1562500G 500 g
B156225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1562500G Supplier TCI America Supplier No. B1562500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2716-10-1
Molecular Formula C24H28N2
Molecular Weight (g/mol) 344.50
MDL Number MFCD00191313
InChI Key HESXPOICBNWMPI-UHFFFAOYSA-N
PubChem CID 75930
IUPAC Name 4-[2-[4-[2-(4-aminophenyl)propan-2-yl]phenyl]propan-2-yl]aniline
SMILES CC(C)(C1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)N)C3=CC=C(C=C3)N

Specifications

Melting Point 165°C
Quantity 500 g
Synonym 1,4-Bis[2-(4-aminophenyl)-2-propyl]benzene, 4,4′-(1,4-Phenylenediisopropylidene)bisaniline, Bisaniline P
Formula Weight 344.50
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha′-Bis(4-aminophenyl)-1,4-diisopropylbenzene
EINECSNumber (4)-1573
RTECSNumber CY1202650
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.