accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOH(E/Z)

CAS RN 2716-53-2

IUPAC Name: 2,3-dihydroxypropyl octadec-9-enoate
Synonyms: 2,3-dihydroxypropyl octadec-9-enoate
Molecular Weight (g/mol): 356.55
Molecular Formula: C21H40O4
InChi Key: RZRNAYUHWVFMIP-UHFFFAOYNA-N
SMILES: CCCCCCCCC=CCCCCCCCC(=O)OCC(O)CO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results