Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monoelaidin, TCI America™
SDP

Catalog No. M10751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10751G Supplier TCI America Supplier No. M10751G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2716-53-2
Molecular Formula C21H40O4
Molecular Weight (g/mol) 356.55
MDL Number MFCD00069635
InChI Key RZRNAYUHWVFMIP-UHFFFAOYNA-N
Synonym Glycerol alpha-Monoelaidate, 1-Mono-trans-9-octadecenoyl Glycerol
PubChem CID 5364833
IUPAC Name 2,3-dihydroxypropyl octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC(O)CO

Specifications

Melting Point 58°C
Color White-Yellow
Quantity 1 g
Formula Weight 356.55
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Monoelaidin
EINECSNumber (2)-0669
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.