accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOOSOOONNNNNN(E)

CAS RN 2718-90-3

IUPAC Name: disodium 5-azido-2-[(1E)-2-(4-azido-2-sulfonatophenyl)ethenyl]benzene-1-sulfonate
Synonyms: disodium 5-azido-2-[(1E)-2-(4-azido-2-sulfonatophenyl)ethenyl]benzenesulfonate
Molecular Weight (g/mol): 466.35
Molecular Formula: C14H8N6Na2O6S2
InChi Key: HYYYTIDWAIQGHI-SEPHDYHBSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=CC=C1\C=C\C1=CC=C(C=C1S([O-])(=O)=O)N=[N+]=[N-])N=[N+]=[N-]
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results