Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium 4,4'-Diazidostilbene-2,2'-disulfonate Tetrahydrate 98.0+%, TCI America™
SDP

Catalog No. D160625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D160625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D160625G Supplier TCI America Supplier No. D160625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2718-90-3
Molecular Formula C14H10N6Na2O6S2
Molecular Weight (g/mol) 468.37
MDL Number MFCD02091646
InChI Key RVHBUGYORUQPGK-UHFFFAOYSA-N
Synonym 4,4′C-Diazidostilbene-2,2′C-disulfonic Acid Disodium Salt
PubChem CID 131851949
IUPAC Name 5-azido-2-[2-(4-azido-2-sulfophenyl)ethenyl]benzenesulfonic acid;sodium
SMILES C1=CC(=C(C=C1N=[N+]=[N-])S(=O)(=O)O)C=CC2=C(C=C(C=C2)N=[N+]=[N-])S(=O)(=O)O.[Na].[Na]

Specifications

Quantity 25 g
UN Number 1325
Formula Weight 466.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 4,4′-Diazidostilbene-2,2′-disulfonate Tetrahydrate
EINECSNumber (4)-0280&(4)-0955
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.