accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27191-09-9

IUPAC Name: 3-methoxyaniline hydrochloride
Synonyms: M-anisidine hydrochloride
Molecular Weight (g/mol): 159.61
Molecular Formula: C7H10ClNO
InChi Key: FNCKZWCTEPKMQV-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=CC(N)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results