Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Anisidine Hydrochloride 99.0+%, TCI America™
SDP

Catalog No. A04885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04885G 5 g
A048825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A04885G Supplier TCI America Supplier No. A04885G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27191-09-9
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.61
MDL Number MFCD00060220
InChI Key FNCKZWCTEPKMQV-UHFFFAOYSA-N
Synonym 3-Methoxyaniline Hydrochloride
PubChem CID 12232010
IUPAC Name 3-methoxyaniline hydrochloride
SMILES Cl.COC1=CC=CC(N)=C1

Specifications

Melting Point 174°C
Quantity 5 g
UN Number 2811
Formula Weight 159.61
Percent Purity ≥99.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material m-Anisidine Hydrochloride
EINECSNumber (3)-0682
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.