accessibility menu, dialog, popup

UserName

Viewing profile as user:
Te

CAS RN 272-35-5

IUPAC Name: 1-benzotellurophene
Synonyms: 1-benzotellurophene
Molecular Weight (g/mol): 229.74
Molecular Formula: C8H6Te
InChi Key: WDKYOIMDQCVVST-UHFFFAOYSA-N
SMILES: [Te]1C=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results