Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[b]tellurophene 96%, Thermo Scientific™

Catalog No. AC276330010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC276330010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC276330010 Supplier Thermo Scientific Chemicals Supplier No. 276330010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 272-35-5
Molecular Formula C8H6Te
Molecular Weight (g/mol) 229.736
InChI Key WDKYOIMDQCVVST-UHFFFAOYSA-N
Synonym benzo b tellurophene, telluranaphthene, #
PubChem CID 136083
IUPAC Name 1-benzotellurophene
SMILES C1=CC=C2C(=C1)C=C[Te]2

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 65°C to 70°C
Color Yellow
Infrared Spectrum Authentic
Quantity 1 g
Formula Weight 229.74
Percent Purity ≥95%
Physical Form Shiny Flakes
Chemical Name or Material Benzo[b]tellurophene, 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.