accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27205-03-4

IUPAC Name: 2-{4-[4-(2-hydroxyethoxy)benzenesulfonyl]phenoxy}ethan-1-ol
Synonyms: 2-{4-[4-(2-hydroxyethoxy)benzenesulfonyl]phenoxy}ethanol
Molecular Weight (g/mol): 338.37
Molecular Formula: C16H18O6S
InChi Key: UTNSTOOXQPHXJQ-UHFFFAOYSA-N
SMILES: OCCOC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(OCCO)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results