accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2721-22-4

IUPAC Name: 6-nonyloxan-2-one
Synonyms: 6-nonyloxan-2-one
Molecular Weight (g/mol): 226.36
Molecular Formula: C14H26O2
InChi Key: SKQYTJLYRIFFCO-UHFFFAOYNA-N
SMILES: CCCCCCCCCC1CCCC(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results