Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta-Tetradecanolactone 98.0+%, TCI America™
SDP

Catalog No. T112925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T112925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T112925ML Supplier TCI America Supplier No. T112925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2721-22-4
Molecular Formula C14H26O2
Molecular Weight (g/mol) 226.36
MDL Number MFCD00059809
InChI Key SKQYTJLYRIFFCO-UHFFFAOYNA-N
Synonym delta-tetradecalactone, 5-tetradecanolide, 2h-pyran-2-one, tetrahydro-6-nonyl, delta-tetradecanolide, delta-tetradecanolactone, tetrahydro-6-nonyl-2h-pyran-2-one, fema no. 3590, 6-nonyltetrahydro-2h-pyran-2-one, delta-myristolactone, 5-hydroxytetradecanoic acid delta-lactone
PubChem CID 520296
IUPAC Name 6-nonyloxan-2-one
SMILES CCCCCCCCCC1CCCC(=O)O1

Specifications

Color Colorless
Boiling Point 175°C
Quantity 25 mL
Formula Weight 226.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material delta-Tetradecanolactone
EINECSNumber (5)-0698
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.