accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONNHNCH3H3CCH3CH3

CAS RN 2725-22-6

IUPAC Name: 6-[4,6-bis(2,4-dimethylphenyl)-2,5-dihydro-1,3,5-triazin-2-ylidene]-2-(octyloxy)cyclohexa-2,4-dien-1-one
Synonyms: 6-[4,6-bis(2,4-dimethylphenyl)-5H-1,3,5-triazin-2-ylidene]-2-(octyloxy)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 509.69
Molecular Formula: C33H39N3O2
InChi Key: DUSCWEXFMYJRHL-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=CC=CC(C1=O)=C1N=C(NC(=N1)C1=CC=C(C)C=C1C)C1=CC=C(C)C=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results