Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine 97.0+%, TCI America™
SDP

Catalog No. B43995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43995G 5 g
B439925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43995G Supplier TCI America Supplier No. B43995G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2725-22-6
Molecular Formula C33H39N3O2
Molecular Weight (g/mol) 509.69
MDL Number MFCD09834146
InChI Key DUSCWEXFMYJRHL-UHFFFAOYSA-N
Synonym 2-[4,6-Bis(2,4-dimethylphenyl)-1,3,5-triazin-2-yl]-5-(n-octyloxy)phenol
PubChem CID 6850826
IUPAC Name 6-[4,6-bis(2,4-dimethylphenyl)-2,5-dihydro-1,3,5-triazin-2-ylidene]-2-(octyloxy)cyclohexa-2,4-dien-1-one
SMILES CCCCCCCCOC1=CC=CC(C1=O)=C1N=C(NC(=N1)C1=CC=C(C)C=C1C)C1=CC=C(C)C=C1C

Specifications

Melting Point 90°C
Color Yellow
Boiling Point 695°C
Quantity 5 g
Formula Weight 509.69
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine
EINECSNumber (5)-6335
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.