accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2727965-45-7

IUPAC Name: (2E)-N-[4-(1-benzoylpiperidin-4-yl)butyl]-3-(pyridin-3-yl)prop-2-enamide hydrochloride
Synonyms: daporinad hydrochloride
Molecular Weight (g/mol): 427.97
Molecular Formula: C24H30ClN3O2
InChi Key: MULSIBUGDPOSHV-CALJPSDSSA-N
SMILES: Cl.O=C(NCCCCC1CCN(CC1)C(=O)C1=CC=CC=C1)\C=C\C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results