accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 273-15-4

IUPAC Name: 2λ⁴,1,3-benzoselenadiazole
Synonyms: 2λ⁴,1,3-benzoselenadiazole
Molecular Weight (g/mol): 183.08
Molecular Formula: C6H4N2Se
InChi Key: AYTPIVIDHMVGSX-UHFFFAOYSA-N
SMILES: C1=CC=C2N=[Se]=NC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results